missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzyl alcohol, 99%

Catalog No. AAA1401614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1401614 25 g
AAA1401622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1401614 Supplier Thermo Scientific Chemicals Supplier No. A1401614
Only null left

CAS: 612-16-8 | C8H10O2 | 138.166 g/mol

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Not miscible or difficult to mix in water. Soluble in ethanol.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong acids.

Chemical Identifiers

CAS 612-16-8
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00004611
InChI Key WYLYBQSHRJMURN-UHFFFAOYSA-N
Synonym 2-methoxybenzyl alcohol, 2-methoxyphenyl methanol, o-methoxybenzyl alcohol, benzenemethanol, 2-methoxy, o-anisyl alcohol, 2-methoxybenzylalcohol, 2-methoxybenzenemethanol, unii-457nq8ngcg, 457nq8ngcg, 2-methoxyphenyl methan-1-ol
PubChem CID 69154
IUPAC Name (2-methoxyphenyl)methanol
SMILES COC1=CC=CC=C1CO

Specifications

Density 1.12
Boiling Point 248°C to 250°C
Flash Point >110°C (230°F)
Refractive Index 1.5475
Quantity 25 g
Beilstein 2043673
Solubility Information Not miscible or difficult to mix in water. Soluble in ethanol.
Formula Weight 138.17
Percent Purity 99%
Chemical Name or Material 2-Methoxybenzyl alcohol
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-296-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only