missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzonitrile 98.0+%, TCI America™

Catalog No. M12805G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12805G 5 g
M128025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12805G Supplier TCI America Supplier No. M12805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6609-56-9
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001783
InChI Key FSTPMFASNVISBU-UHFFFAOYSA-N
Synonym 2-cyanoanisole, o-methoxybenzonitrile, 2-methoxy-benzonitrile, benzonitrile, 2-methoxy, 2-methoxybenzenecarbonitrile, o-anisolnitrile, o-anisonitrile, methoxybenzonitrile, 2-methxybenzonitrile, o-methyoxybenzonitrile
PubChem CID 81086
IUPAC Name 2-methoxybenzonitrile
SMILES COC1=CC=CC=C1C#N

Specifications

Color White-Yellow
Boiling Point 256°C
Quantity 5 g
UN Number 2811
Formula Weight 133.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methoxybenzonitrile