missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzonitrile, 98%

Catalog No. AAL0129306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0129306 5 g
AAL0129314 25 g
AAL0129322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0129306 Supplier Thermo Scientific Chemicals Supplier No. L0129306

CAS: 6609-56-9 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6609-56-9
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00001783
InChI Key FSTPMFASNVISBU-UHFFFAOYSA-N
Synonym 2-cyanoanisole, o-methoxybenzonitrile, 2-methoxy-benzonitrile, benzonitrile, 2-methoxy, 2-methoxybenzenecarbonitrile, o-anisolnitrile, o-anisonitrile, methoxybenzonitrile, 2-methxybenzonitrile, o-methyoxybenzonitrile
PubChem CID 81086
IUPAC Name 2-methoxybenzonitrile
SMILES COC1=CC=CC=C1C#N

Specifications

Density 1.093
Boiling Point 256°C
Flash Point >110°C (230°F)
Odor Almond-like
Refractive Index 1.5465
Quantity 5 g
UN Number UN3276
Beilstein 2207134
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material 2-Methoxybenzonitrile
Show More Show Less
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
EINECSNumber 229-559-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only