missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzoic acid, 98+%

Catalog No. AAA1351336
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1351336 500 g
AAA1351322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1351336 Supplier Thermo Scientific Chemicals Supplier No. A1351336
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 579-75-9 | C8H8O3 | 152.149 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 579-75-9
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.149
MDL Number MFCD00002431
InChI Key ILUJQPXNXACGAN-UHFFFAOYSA-N
Synonym o-anisic acid, o-methoxybenzoic acid, 2-anisic acid, o-methylsalicylic acid, benzoic acid, 2-methoxy, salicylic acid methyl ether, 2-methoxy-benzoic acid, 2-methoxybenzoicacid, o-methoxy benzoic acid, ortho-methoxybenzoic acid
PubChem CID 11370
ChEBI CHEBI:421840
IUPAC Name 2-methoxybenzoic acid
SMILES COC1=CC=CC=C1C(=O)O

Specifications

Melting Point 99°C to 103°C
Odor Odorless
Quantity 500 g
Beilstein 509929
Formula Weight 152.15
Percent Purity ≥98%
Chemical Name or Material 2-Methoxybenzoic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-447-8
RTECSNumber BZ4385000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only