missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzeneboronic acid, 97%

CAS: 5720-06-9 | C7H9BO3 | 151.96 g/mol

Supplier:  Thermo Scientific Chemicals B2107114

 View more versions of this product

Catalog No. AAB2107114


Only null left
Explore available promotions

Description

Description

2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications

Specifications

2-Methoxybenzeneboronic acid
102°C to 106°C
MFCD00236047
3030981
Insoluble in water.
COC1=CC=CC=C1B(O)O
151.96
151.96
5720-06-9
C7H9BO3
25 g
2-methoxybenzeneboronic acid, 2-methoxyphenyl boronic acid, o-methoxyphenylboronic acid, 2-methoxyphenyl boranediol, 2-boronoanisole, 2-methoxypenylboronic acid, o-methoxyphenyl boronic acid, 2-methoxy phenylboronic acid, 2-methoxybenzene boronic acid
ROEQGIFOWRQYHD-UHFFFAOYSA-N
(2-methoxyphenyl)boronic acid
2733958
97%

Chemical Identifiers

5720-06-9
151.96
ROEQGIFOWRQYHD-UHFFFAOYSA-N
2733958
COC1=CC=CC=C1B(O)O
C7H9BO3
MFCD00236047
2-methoxybenzeneboronic acid, 2-methoxyphenyl boronic acid, o-methoxyphenylboronic acid, 2-methoxyphenyl boranediol, 2-boronoanisole, 2-methoxypenylboronic acid, o-methoxyphenyl boronic acid, 2-methoxy phenylboronic acid, 2-methoxybenzene boronic acid
(2-methoxyphenyl)boronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only