missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxybenzeneboronic acid, 97%

Catalog No. p-7025539
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Catalog No. Quantity
AAB2107103 1 g
AAB2107106 5 g
AAB2107114 25 g
3 options

Catalog No. AAB2107103

Supplier: Thermo Scientific Chemicals B2107103

Only null left

CAS: 5720-06-9 | C7H9BO3 | 151.96 g/mol

2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxylphenylboronic acid is a phenylboronic acid used to investigate boron function in plants. It is used in Suzuki reaction, organic reaction that is classified as a coupling reaction where the coupling partners are a boronic acid with a halide catalyzed by a palladium(0) complex.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5720-06-9
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD00236047
InChI Key ROEQGIFOWRQYHD-UHFFFAOYSA-N
Synonym 2-methoxybenzeneboronic acid, 2-methoxyphenyl boronic acid, o-methoxyphenylboronic acid, 2-methoxyphenyl boranediol, 2-boronoanisole, 2-methoxypenylboronic acid, o-methoxyphenyl boronic acid, 2-methoxy phenylboronic acid, 2-methoxybenzene boronic acid
PubChem CID 2733958
IUPAC Name (2-methoxyphenyl)boronic acid
SMILES COC1=CC=CC=C1B(O)O

Specifications

Melting Point 102°C to 106°C
Quantity 1 g
Beilstein 3030981
Solubility Information Insoluble in water.
Formula Weight 151.96
Percent Purity 97%
Chemical Name or Material 2-Methoxybenzeneboronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only