missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxyacetophenone, 99%

Catalog No. AC125670250
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC125670250 25 mL Glass bottle
AC125671000 100 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC125670250 Supplier Thermo Scientific Chemicals Supplier No. 125670250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 579-74-8 | C9H10O2 | 150.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 579-74-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008725
InChI Key DWPLEOPKBWNPQV-UHFFFAOYSA-N
Synonym 2'-methoxyacetophenone, 1-2-methoxyphenyl ethanone, o-methoxyacetophenone, ethanone, 1-2-methoxyphenyl, o-acetylanisole, 1-2-methoxyphenyl ethan-1-one, o-acetanisole, 2-acetylanisole, 2-methyoxyacetophenone oxime, ortho-methoxyacetophenone
PubChem CID 68481
IUPAC Name 1-(2-methoxyphenyl)ethanone
SMILES COC1=CC=CC=C1C(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown or Orange to Yellow
Density 1.0900g/mL
Boiling Point 131.0°C (18.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5383 to 1.5403
Linear Formula CH3OC6H4COCH3
Quantity 25 mL
Beilstein 08, 85
Specific Gravity 1.09
Solubility Information Solubility in water: immiscible
Formula Weight 150.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2'-Methoxyacetophenone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-446-2
TSCA TSCA

RUO – Research Use Only