missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-methylaniline 98.0+%, TCI America™

Catalog No. p-7132861
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
M18045G 5 g
M180425G 25 g
2 options

Catalog No. M18045G

Supplier: TCI America M18045G

Only null left

Chemical Identifiers

CAS 50868-73-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00075455
InChI Key HKOJYPPTIPJZAZ-UHFFFAOYSA-N
Synonym 6-Methoxy-o-toluidine, 2-Amino-1-methoxy-3-methylbenzene
PubChem CID 459249
IUPAC Name 2-methoxy-6-methylaniline
SMILES COC1=CC=CC(C)=C1N

Specifications

Color Brown-White
Boiling Point 116°C
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Methoxy-6-methylaniline