missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethyl)benzaldehyde, 98%, Thermo Scientific Chemicals

Catalog No. p-7028729
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2613503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2613503 Supplier Thermo Scientific Chemicals Supplier No. H2613503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 146539-83-5 | Molecular Formula: C9H7F3O2 | Molecular Weight: 204.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 146539-83-5
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD03788528
InChI Key WFRURKYDETYZGF-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethyl benzaldehyde, 2-methoxy-5-trifluoromethyl-benzaldehyde, benzaldehyde, 2-methoxy-5-trifluoromethyl, 5-trifluoromethyl-2-methoxybenzaldehyde, pubchem8509, acmc-209cx7, ksc496k3j, benzaldehyde,2-methoxy-5-trifluoromethyl
PubChem CID 2779272
IUPAC Name 2-methoxy-5-(trifluoromethyl)benzaldehyde
SMILES COC1=CC=C(C=C1C=O)C(F)(F)F

Specifications

Melting Point 85°C to 89°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 204.15
Percent Purity 98%
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)benzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only