missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethoxy)benzaldehyde 98.0+%, TCI America™

Catalog No. M19455G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M19455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M19455G Supplier TCI America Supplier No. M19455G
Only null left

Specifications

CAS 145742-65-0
Color Yellow
Molecular Formula C9H7F3O3
Quantity 5 g
MDL Number MFCD04115958
Synonym 2-methoxy-5-trifluoromethoxy benzaldehyde, 2-formyl-4-trifluoromethoxy anisole, 2-methoxy-5-trifluoromethoxy-benzaldehyde, acmc-209cvk, 2-methoxy-5-trifluoromethoxybenzaidehyde, 2-methoxy-5-trifluoromethyloxy benzaldehyde, benzaldehyde,2-methoxy-5-trifluoromethoxy, benzaldehyde, 2-methoxy-5-trifluoromethoxy
InChI Key ATRDCTRZAJKDPL-UHFFFAOYSA-N
SMILES COC1=CC=C(OC(F)(F)F)C=C1C=O
IUPAC Name 2-methoxy-5-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol) 220.15
PubChem CID 2783364
Formula Weight 220.15
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-5-(trifluoromethoxy)benzaldehyde
Show More Show Less