missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-methylphenyl isocyanate, 97%

Catalog No. AAL1600106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1600106 5 g
AAL1600103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1600106 Supplier Thermo Scientific Chemicals Supplier No. L1600106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59741-04-7 | C9H9NO2 | 163.176 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59741-04-7
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.176
MDL Number MFCD00013869
InChI Key IDOHLSFNBKNRRJ-UHFFFAOYSA-N
Synonym 2-methoxy-5-methylphenyl isocyanate, 2-methoxy-5-methylphenylisocyanate, 2-isocyanato-1-methoxy-4-methyl-benzene, benzene, 2-isocyanato-1-methoxy-4-methyl, acmc-20anrw, 2-methoxy-5-methylbenzenisocyanate, 2-methoxy-5-methylpheny lisocyanate, 2-isocyanato-1-methyloxy-4-methylbenzene, benzene, 2-isocyanato-1-methoxy-4-methyl-9ci
PubChem CID 4141520
IUPAC Name 2-isocyanato-1-methoxy-4-methylbenzene
SMILES CC1=CC(=C(C=C1)OC)N=C=O

Specifications

Density 1.112
Boiling Point 240°C
Flash Point >110°C (230°F)
Refractive Index 1.537
Quantity 5 g
UN Number UN2206
Beilstein 3604599
Sensitivity Moisture sensitive
Formula Weight 163.18
Percent Purity 97%
Chemical Name or Material 2-Methoxy-5-methylphenyl isocyanate
Show More Show Less
Hazard Category H302+H312+H332-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H331-H334-H302-H315-H319-H335
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P342+P311-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only