missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™

Catalog No. p-7133347
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26501G 1 g
M26505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26501G Supplier TCI America Supplier No. M26501G

Chemical Identifiers

CAS 445264-61-9
Molecular Formula C12H18BNO3
Molecular Weight (g/mol) 235.09
MDL Number MFCD05663858
InChI Key QOGNDJLSYMJGPP-UHFFFAOYSA-N
Synonym 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxyl-5-pyridineboronic acid pinacol ester, 2-methoxypyridine-5-boronic acid pinacol ester, 6-methoxypyridine-3-boronic acid pinacol ester, 2-methoxy-5-pyridineboronic acid pinacol ester, 6-methoxypyridin-3-ylboronic acid pinacol ester, pyridine, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methoxypyridine-5-boronic acid, pinacol ester, 2-methoxypyridin-5-yl boronic acid pinacol ester, 2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
PubChem CID 16217714
IUPAC Name 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OC

Specifications

Melting Point 53°C
Color White
Boiling Point 102°C
Quantity 1 g
Formula Weight 235.09
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine