accessibility menu, dialog, popup

UserName

CH3ONBOH3CH3CH3CCH3O

CAS RN 445264-61-9

IUPAC Name: 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 235.09
Molecular Formula: C12H18BNO3
InChi Key: QOGNDJLSYMJGPP-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=N1)B1OC(C)(C)C(C)(C)O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results