missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrotoluene 98.0+%, TCI America™

Catalog No. p-7132805
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M168125G 25 g
M1681500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M168125G Supplier TCI America Supplier No. M168125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 13120-77-9
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
MDL Number MFCD00043912
InChI Key WVQGZNRUEVFXKR-UHFFFAOYSA-N
Synonym 1-Methoxy-2-methyl-5-nitrobenzene, 2-Methyl-5-nitroanisole
PubChem CID 83159
IUPAC Name 2-methoxy-1-methyl-4-nitrobenzene
SMILES COC1=CC(=CC=C1C)[N+]([O-])=O

Specifications

Melting Point 73°C
Color Yellow
Quantity 25 g
Formula Weight 167.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-4-nitrotoluene
EINECSNumber (3)-0799
TSCA Yes