missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrobenzonitrile, 98%, Thermo Scientific™

 

Supplier:  Thermo Scientific Chemicals 440220050

Catalog No. AC440220050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

 

Specifications

Specifications

2-Methoxy-4-nitrobenzonitrile
97.5
Authentic
Glass bottle
5g
MLIKCKXLGYEGAO-UHFFFAOYSA-N
2-methoxy-4-nitrobenzonitrile
291867
98%
101084-96-2
100.0
97.5% min. (HPLC)
C8H6N2O3
2-cyano-5-nitroanisole, acmc-20a4y1, 4-nitro-2-methoxybenzonitrile, 2-methoxy-4-nitro-benzonitrile, 2-methyloxy-4-nitrobenzonitrile, 2-methoxy-4-nitrobenzonitrile
COC1=C(C=CC(=C1)[N+](=O)[O-])C#N
178.15
178.15

Chemical Identifiers

101084-96-2
178.15
2-cyano-5-nitroanisole, acmc-20a4y1, 4-nitro-2-methoxybenzonitrile, 2-methoxy-4-nitro-benzonitrile, 2-methyloxy-4-nitrobenzonitrile, 2-methoxy-4-nitrobenzonitrile
2-methoxy-4-nitrobenzonitrile
C8H6N2O3
MLIKCKXLGYEGAO-UHFFFAOYSA-N
291867
COC1=C(C=CC(=C1)[N+](=O)[O-])C#N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only