missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrobenzoic acid, 98%

Catalog No. AAB2552306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2552306 5 g
AAB2552303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2552306 Supplier Thermo Scientific Chemicals Supplier No. B2552306
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2597-56-0 | C8H7NO5 | 197.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2597-56-0
Molecular Formula C8H7NO5
Molecular Weight (g/mol) 197.15
MDL Number MFCD00216573
InChI Key KPJXEWJRJKEOCD-UHFFFAOYSA-N
Synonym 2-methoxy-4-nitrobenzoic acid, 4-nitro-o-anisic acid, 4-nitro-2-methoxybenzoic acid, rarechem al bo 1464, 2-carboxy-5-nitroanisole, 2-methoxy-4-nitro-benzoic acid, benzoic acid, 2-methoxy-4-nitro, 4-nitro-2-methoxybenzoicacid, pubchem4977, acmc-20a5eb
PubChem CID 75776
IUPAC Name 2-methoxy-4-nitrobenzoic acid
SMILES COC1=CC(=CC=C1C(O)=O)[N+]([O-])=O

Specifications

Melting Point 145°C to 148°C
Quantity 5 g
Formula Weight 197.15
Percent Purity 98%
Chemical Name or Material 2-Methoxy-4-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-998-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only