missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7133206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24031G 1 g
M24035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24031G Supplier TCI America Supplier No. M24031G
Only null left

Chemical Identifiers

CAS 21320-91-2
Molecular Formula C7H6ClNO5S
Molecular Weight (g/mol) 251.64
MDL Number MFCD03094697
InChI Key QECYXMKYZQXEHM-UHFFFAOYSA-N
Synonym 2-methoxy-4-nitrobenzene-1-sulfonyl chloride, 2-methoxy-4-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 2-methoxy-4-nitro, pubchem11714, acmc-1ck5c, ksc207c7t, 2-chlorosulphonyl-5-nitroanisole, chloro 2-methoxy-4-nitrophenyl sulfone, 2-methoxy-4-nitrobenzensulfonyl chloride, 2-methoxy-4-nitrophenylsulfonyl chloride
PubChem CID 309458
IUPAC Name 2-methoxy-4-nitrobenzene-1-sulfonyl chloride
SMILES COC1=CC(=CC=C1S(Cl)(=O)=O)[N+]([O-])=O

Specifications

Melting Point 99°C
Color Yellow
Quantity 1 g
UN Number 3261
Formula Weight 251.64
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-4-nitrobenzenesulfonyl Chloride