missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrobenzenesulfonyl chloride, 97%

Catalog No. p-7047897
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1990203 1 g
AAL1990206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1990203 Supplier Thermo Scientific Chemicals Supplier No. L1990203
Only null left

CAS: 21320-91-2 | C7H6ClNO5S | 251.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 21320-91-2
Molecular Formula C7H6ClNO5S
Molecular Weight (g/mol) 251.64
MDL Number MFCD03094697
InChI Key QECYXMKYZQXEHM-UHFFFAOYSA-N
Synonym 2-methoxy-4-nitrobenzene-1-sulfonyl chloride, 2-methoxy-4-nitrobenzenesulfonylchloride, benzenesulfonyl chloride, 2-methoxy-4-nitro, pubchem11714, acmc-1ck5c, ksc207c7t, 2-chlorosulphonyl-5-nitroanisole, chloro 2-methoxy-4-nitrophenyl sulfone, 2-methoxy-4-nitrobenzensulfonyl chloride, 2-methoxy-4-nitrophenylsulfonyl chloride
PubChem CID 309458
SMILES COC1=CC(=CC=C1S(Cl)(=O)=O)[N+]([O-])=O

Specifications

Melting Point 96°C to 99°C
Quantity 1 g
UN Number UN3261
Beilstein 2136023
Sensitivity Moisture sensitive
IUPAC Name 2-methoxy-4-nitrobenzene-1-sulfonyl chloride
Formula Weight 251.65
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-nitrobenzenesulfonyl chloride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only