missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-4-nitrobenzaldehyde 98.0+%, TCI America™
$145.40 - $495.84
Chemical Identifiers
| CAS | 136507-15-8 |
|---|---|
| Molecular Formula | C8H7NO4 |
| Molecular Weight (g/mol) | 181.147 |
| MDL Number | MFCD02683052 |
| InChI Key | LEBUUZXTHMCZQZ-UHFFFAOYSA-N |
| PubChem CID | 2759599 |
| IUPAC Name | 2-methoxy-4-nitrobenzaldehyde |
| SMILES | COC1=C(C=CC(=C1)[N+](=O)[O-])C=O |
Chemical Identifiers
| 136507-15-8 | |
| 181.147 | |
| LEBUUZXTHMCZQZ-UHFFFAOYSA-N | |
| 2-methoxy-4-nitrobenzaldehyde |
| C8H7NO4 | |
| MFCD02683052 | |
| 2759599 | |
| COC1=C(C=CC(=C1)[N+](=O)[O-])C=O |
Specifications
| 136507-15-8 | |
| Yellow | |
| MFCD02683052 | |
| LEBUUZXTHMCZQZ-UHFFFAOYSA-N | |
| 2-methoxy-4-nitrobenzaldehyde | |
| 2759599 | |
| ≥98.0% (GC) | |
| 2-Methoxy-4-nitrobenzaldehyde |
| 122°C | |
| C8H7NO4 | |
| 1 g | |
| COC1=C(C=CC(=C1)[N+](=O)[O-])C=O | |
| 181.147 | |
| 181.15 | |
| Crystalline Powder |
Safety and Handling
TSCA : No