missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitrobenzaldehyde 98.0+%, TCI America™

Catalog No. M26565G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26565G 5 g
M26561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26565G Supplier TCI America Supplier No. M26565G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 136507-15-8
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.147
MDL Number MFCD02683052
InChI Key LEBUUZXTHMCZQZ-UHFFFAOYSA-N
PubChem CID 2759599
IUPAC Name 2-methoxy-4-nitrobenzaldehyde
SMILES COC1=C(C=CC(=C1)[N+](=O)[O-])C=O

Specifications

Melting Point 122°C
Color Yellow
Quantity 5 g
Formula Weight 181.15
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-4-nitrobenzaldehyde