missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitroaniline 99.0+%, TCI America™

Catalog No. p-7132001
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M011825G 25 g
M0118500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M011825G Supplier TCI America Supplier No. M011825G
Only null left

Chemical Identifiers

CAS 97-52-9
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00007363
InChI Key GVBHRNIWBGTNQA-UHFFFAOYSA-N
Synonym fast red b base, benzenamine, 2-methoxy-4-nitro, 4-nitro-o-anisidine, fast red base b, fast red b, red b base, diazo fast red b, 2-amino-5-nitroanisole, azoamine pink o, fast red 5na base
PubChem CID 7337
IUPAC Name 2-methoxy-4-nitroaniline
SMILES COC1=CC(=CC=C1N)[N+]([O-])=O

Specifications

Melting Point 140°C
Quantity 25 g
Formula Weight 168.15
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-4-nitroaniline
EINECSNumber (3)-0803
RTECSNumber BZ7170000
TSCA Yes