missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-nitroaniline, 98%

Catalog No. p-3758314
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150640050 5 g Glass bottle
AC150641000 100 g Plastic bottle
AC150645000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC150640050 Supplier Thermo Scientific Chemicals Supplier No. 150640050
Only null left

CAS: 97-52-9 | C7H8N2O3 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 97-52-9
Molecular Formula C7H8N2O3
Molecular Weight (g/mol) 168.15
MDL Number MFCD00007363
InChI Key GVBHRNIWBGTNQA-UHFFFAOYSA-N
Synonym fast red b base, benzenamine, 2-methoxy-4-nitro, 4-nitro-o-anisidine, fast red base b, fast red b, red b base, diazo fast red b, 2-amino-5-nitroanisole, azoamine pink o, fast red 5na base
PubChem CID 7337
IUPAC Name 2-methoxy-4-nitroaniline
SMILES COC1=CC(=CC=C1N)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 139.0°C to 142.0°C
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3OC6H3(NO2)NH2
Quantity 5 g
Beilstein 13, 390
Solubility Information Solubility in water: slightly soluble
Formula Weight 168.15
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-4-nitroaniline
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 202-588-6

RUO – Research Use Only