missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-4-methylphenol, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 2.0°C |
| Color | Yellow |
| Density | 1.0900g/mL |
| Boiling Point | 221.0°C |
| Flash Point | 90°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98.5% min. (GC) |
| Packaging | Glass bottle |
| Show More |
Safety and Handling
| missing translation for 'healthHazard1' | GHS Signal Word: Warning |
| missing translation for 'healthHazard2' | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
| missing translation for 'healthHazard3' | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/e |
| EINECSNumber | 202-252-9 |
| RTECSNumber | GP1755000 |
| TSCA | TSCA |
RUO – Research Use Only