missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methylphenol, 98+%

Catalog No. AAA1360330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1360330 250 g
AAA1360309 10 g
AAA1360318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1360330 Supplier Thermo Scientific Chemicals Supplier No. A1360330

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-51-6 | C8H10O2 | 138.166 g/mol

2-Methoxy-4-methylphenol is used in preparation of renewable bis(cyanate) esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxy-4-methylphenol is used in preparation of renewable bis(cyanate) esters.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 93-51-6
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.166
MDL Number MFCD00002378
InChI Key PETRWTHZSKVLRE-UHFFFAOYSA-N
Synonym creosol, 4-methylguaiacol, 2-methoxy-p-cresol, homoguaiacol, p-methylguaiacol, phenol, 2-methoxy-4-methyl, p-creosol, 4-methyl guaiacol, 2-methoxy-4-cresol, 4-hydroxy-3-methoxytoluene
PubChem CID 7144
IUPAC Name 2-methoxy-4-methylphenol
SMILES CC1=CC(=C(C=C1)O)OC

Specifications

Melting Point 5°C to 6°C
Density 1.092
Boiling Point 220°C to 222°C
Flash Point 99°C (210°F)
Odor Characteristic
Refractive Index 1.537
Quantity 250 g
Beilstein 1862340
Merck Index 14,2571
Solubility Information Slightly soluble in water.
Formula Weight 138.17
Percent Purity ≥98%
Chemical Name or Material 2-Methoxy-4-methylphenol
Show More Show Less
Hazard Category H302-H315-H317-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c
EINECSNumber 202-252-9
RTECSNumber GP1755000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only