missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-methylbenzenesulfonamide, 95%

Catalog No. AAL1694703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1694703 1 g
AAL1694706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1694703 Supplier Thermo Scientific Chemicals Supplier No. L1694703
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 59554-39-1 | C8H11NO3S | 201.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 59554-39-1
Molecular Formula C8H11NO3S
Molecular Weight (g/mol) 201.24
MDL Number MFCD01318156
InChI Key GQUFSEOISDLWMN-UHFFFAOYSA-N
Synonym 2-methoxy-4-methyl-benzenesulfonamide, 2-methoxy-4-methylbenzene-1-sulfonamide, acmc-20ans2, 3-methoxy-p-toluenesulfonamide, benzenesulfonamide,2-methoxy-4-methyl, 2-methoxy-4-methyl-1-benzenesulfonamide
PubChem CID 5093369
IUPAC Name 2-methoxy-4-methylbenzenesulfonamide
SMILES CC1=CC(=C(C=C1)S(=O)(=O)N)OC

Specifications

Melting Point 168°C to 170°C
Quantity 1 g
Formula Weight 201.25
Percent Purity 95%
Chemical Name or Material 2-Methoxy-4-methylbenzenesulfonamide
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only