missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-3-nitropyridine 98.0+%, TCI America™

Catalog No. M13955G
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
M13955G 5 g
1 options

Catalog No. M13955G

Supplier: TCI America M13955G

Only null left

Chemical Identifiers

CAS 20265-35-4
Molecular Formula C6H6N2O3
Molecular Weight (g/mol) 154.125
MDL Number MFCD00023459
InChI Key WZNQCVOSOCGWJG-UHFFFAOYSA-N
Synonym 2-methoxy-3-nitro-pyridine, pyridine, 2-methoxy-3-nitro, 2-methoxy-3-nitropyridin, pubchem6593, maybridge1_008636, acmc-1cdn9, 2-methoxy-3-nitropyridine?, ksc494o8j, 2-methoxy-3-nitropyridine, 2-methoxy-3-nitropyridine 1g
PubChem CID 253219
IUPAC Name 2-methoxy-3-nitropyridine
SMILES COC1=C(C=CC=N1)[N+](=O)[O-]

Specifications

Melting Point 56°C
Color Yellow
Quantity 5 g
Formula Weight 154.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-3-nitropyridine