missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile 98.0+%, TCI America™

Catalog No. M11635G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11635G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M11635G Supplier TCI America Supplier No. M11635G

Chemical Identifiers

CAS 80866-87-1
Molecular Formula C10H8F3NO
Molecular Weight (g/mol) 215.175
MDL Number MFCD00001844
InChI Key UBOJBAYKXZRZHI-UHFFFAOYSA-N
Synonym 2-methoxy-2-phenyl-3,3,3-trifluoropropionitrile, alpha-methoxy-alpha-trifluoromethyl phenylacetonitrile, methoxyphenyl trifluoromethyl acetonitrile, acmc-1bl9n, -a-methoxy-a-trifluoromethylphenylacetonitrile, +/--a-methoxy-a-trifluoromethylphenylacetonitrile, #, +/--2-methoxy-2-trifluoromethyl phenylacetonitrile, +\--2-methoxy-2-phenyl-3,3,3-trifluoropropionitrile, alpha-trifluoromethyl-alpha-methoxybenzeneacetonitrile
PubChem CID 589652
IUPAC Name 3,3,3-trifluoro-2-methoxy-2-phenylpropanenitrile
SMILES COC(C#N)(C1=CC=CC=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 82°C
Quantity 5 g
Formula Weight 215.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Methoxy-2-phenyl-3,3,3-trifluoropropionitrile