missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-2,4-diphenyl-3(2H)-furanone 98.0+%, TCI America™

Catalog No. p-7132320
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M0722100MG 100 mg
M07221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M0722100MG Supplier TCI America Supplier No. M0722100MG
Only null left

Chemical Identifiers

CAS 50632-57-0
Molecular Formula C17H14O3
Molecular Weight (g/mol) 266.296
MDL Number MFCD00036834
InChI Key BLWINLJDTOJSRU-UHFFFAOYSA-N
PubChem CID 170874
IUPAC Name 2-methoxy-2,4-diphenylfuran-3-one
SMILES COC1(C(=O)C(=CO1)C2=CC=CC=C2)C3=CC=CC=C3

Specifications

Melting Point 96°C
Color White
Quantity 100 mg
Formula Weight 266.30
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-2,4-diphenyl-3(2H)-furanone