accessibility menu, dialog, popup

UserName

CAS RN 50632-57-0

H3COOO

CAS RN 50632-57-0

IUPAC Name: 2-methoxy-2,4-diphenyl-2,3-dihydrofuran-3-one
Synonyms: 2-methoxy-2,4-diphenylfuran-3-one
Molecular Weight (g/mol): 266.3
Molecular Formula: C17H14O3
InChi Key: BLWINLJDTOJSRU-UHFFFAOYNA-N
SMILES: COC1(OC=C(C1=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results