missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-naphthoic Acid 98.0+%, TCI America™

Catalog No. p-7132718
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14175G Supplier TCI America Supplier No. M14175G

Chemical Identifiers

CAS 947-62-6
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00021532
InChI Key OSTYZAHQVPMQHI-UHFFFAOYSA-N
Synonym 2-methoxy-1-naphthoic acid, 2-methoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-methoxy, 1-naphthoic acid, 2-methoxy, acmc-209rtt, 2-methoxy-1-naphtoic acid, 2-methoxy-1-naphthoic acid #, 2-methoxy-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 2-methoxy
PubChem CID 70356
IUPAC Name 2-methoxynaphthalene-1-carboxylic acid
SMILES COC1=C(C2=CC=CC=C2C=C1)C(=O)O

Specifications

Melting Point 178°C
Quantity 5 g
Formula Weight 202.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxy-1-naphthoic Acid