missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-naphthoic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6043403

Catalog No. AAH6043403


Only null left
Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Methoxy-1-naphthoic acid
177°C to 180°C
MFCD00021532
2-methoxy-1-naphthoic acid, 2-methoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-methoxy, 1-naphthoic acid, 2-methoxy, acmc-209rtt, 2-methoxy-1-naphtoic acid, 2-methoxy-1-naphthoic acid #, 2-methoxy-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 2-methoxy
COC1=C(C2=CC=CC=C2C=C1)C(=O)O
202.209
202.21
Solid
947-62-6
C12H10O3
1 g
OSTYZAHQVPMQHI-UHFFFAOYSA-N
2-methoxynaphthalene-1-carboxylic acid
70356
98%

Chemical Identifiers

947-62-6
202.209
OSTYZAHQVPMQHI-UHFFFAOYSA-N
70356
COC1=C(C2=CC=CC=C2C=C1)C(=O)O
C12H10O3
MFCD00021532
2-methoxy-1-naphthoic acid, 2-methoxynaphthoic acid, 1-naphthalenecarboxylic acid, 2-methoxy, 1-naphthoic acid, 2-methoxy, acmc-209rtt, 2-methoxy-1-naphtoic acid, 2-methoxy-1-naphthoic acid #, 2-methoxy-1-naphthalenecarboxylic acid, 1-naphthalenecarboxylicacid, 2-methoxy
2-methoxynaphthalene-1-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-429-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions