missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-1-naphthalenemethanol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 103°C |
| Quantity | 5 g |
| Synonym | 2-methoxy-1-naphthalenemethanol, 2-methoxynaphthalen-1-yl methanol, 1-naphthalenemethanol, 2-methoxy, 1-hydroxymethyl-2-methoxynaphthalene, acmc-1ald1, 2-methoxynaphth-1-yl methanol, 2-methoxynaphthyl methan-1-ol, 1-naphthalenemethanol,2-methoxy |
| Formula Weight | 188.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2-Methoxy-1-naphthalenemethanol |
Safety and Handling
| TSCA | No |