accessibility menu, dialog, popup

UserName

CH3OHO

CAS RN 40696-22-8

IUPAC Name: (2-methoxynaphthalen-1-yl)methanol
Synonyms: (2-methoxynaphthalen-1-yl)methanol
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: VBHARLNUEDIKQD-UHFFFAOYSA-N
SMILES: COC1=CC=C2C=CC=CC2=C1CO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results