missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-naphthaldehyde 98.0+%, TCI America™

$71.23 - $240.64

Chemical Identifiers

CAS 5392-12-1
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004004
InChI Key YIQGLTKAOHRZOL-UHFFFAOYSA-N
Synonym 2-methoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-methoxy, 2-methoxy-1-naphthalenecarboxaldehyde, unii-7q7tx7tb9e, 7q7tx7tb9e, 2-methoxy-1-naphthylaldehyde, 2-methoxy-naphthalene-1-carbaldehyde, 2-methoxynaphthalenecarbaldehyde, 2-methoxy-1-naphthaldehyd, 2-methoxynaphthaldehyde
PubChem CID 79352
IUPAC Name 2-methoxynaphthalene-1-carbaldehyde
SMILES COC1=C(C2=CC=CC=C2C=C1)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10405G
View Documents
TCI America
M10405G
5 g
Each for $71.23
Only null left
 
M104025G
View Documents
TCI America
M104025G
25 g
Each for $240.64
Only null left
 
Description

Description

Specifications

Chemical Identifiers

5392-12-1
186.21
YIQGLTKAOHRZOL-UHFFFAOYSA-N
79352
COC1=C(C2=CC=CC=C2C=C1)C=O
C12H10O2
MFCD00004004
2-methoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-methoxy, 2-methoxy-1-naphthalenecarboxaldehyde, unii-7q7tx7tb9e, 7q7tx7tb9e, 2-methoxy-1-naphthylaldehyde, 2-methoxy-naphthalene-1-carbaldehyde, 2-methoxynaphthalenecarbaldehyde, 2-methoxy-1-naphthaldehyd, 2-methoxynaphthaldehyde
2-methoxynaphthalene-1-carbaldehyde

Specifications

5392-12-1
205°C
MFCD00004004
2-methoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-methoxy, 2-methoxy-1-naphthalenecarboxaldehyde, unii-7q7tx7tb9e, 7q7tx7tb9e, 2-methoxy-1-naphthylaldehyde, 2-methoxy-naphthalene-1-carbaldehyde, 2-methoxynaphthalenecarbaldehyde, 2-methoxy-1-naphthaldehyd, 2-methoxynaphthaldehyde
COC1=C(C2=CC=CC=C2C=C1)C=O
186.21
186.21
Crystalline Powder
84°C
C12H10O2
5 g
YIQGLTKAOHRZOL-UHFFFAOYSA-N
2-methoxynaphthalene-1-carbaldehyde
79352
≥98.0% (GC)
2-Methoxy-1-naphthaldehyde
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents