Learn More
2-Methoxy-1-naphthaldehyde 98.0+%, TCI America™
Supplier:  TCI America M104025G
 View more versions of this product
Catalog No. M104025G
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
2-Methoxy-1-naphthaldehyde
84°C
C12H10O2
25 g
YIQGLTKAOHRZOL-UHFFFAOYSA-N
2-methoxynaphthalene-1-carbaldehyde
79352
≥98.0% (GC)
5392-12-1
205°C
MFCD00004004
2-methoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-methoxy, 2-methoxy-1-naphthalenecarboxaldehyde, unii-7q7tx7tb9e, 7q7tx7tb9e, 2-methoxy-1-naphthylaldehyde, 2-methoxy-naphthalene-1-carbaldehyde, 2-methoxynaphthalenecarbaldehyde, 2-methoxy-1-naphthaldehyd, 2-methoxynaphthaldehyde
COC1=C(C2=CC=CC=C2C=C1)C=O
186.21
186.21
Crystalline Powder
Chemical Identifiers
5392-12-1
186.21
YIQGLTKAOHRZOL-UHFFFAOYSA-N
79352
COC1=C(C2=CC=CC=C2C=C1)C=O
C12H10O2
MFCD00004004
2-methoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-methoxy, 2-methoxy-1-naphthalenecarboxaldehyde, unii-7q7tx7tb9e, 7q7tx7tb9e, 2-methoxy-1-naphthylaldehyde, 2-methoxy-naphthalene-1-carbaldehyde, 2-methoxynaphthalenecarbaldehyde, 2-methoxy-1-naphthaldehyd, 2-methoxynaphthaldehyde
2-methoxynaphthalene-1-carbaldehyde
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
TSCA : No