missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-naphthaldehyde, 99%

CAS: 5392-12-1 | C12H10O2 | 186.21 g/mol

$46.42 - $129.92

Chemical Identifiers

CAS 5392-12-1
MDL Number MFCD00004004
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2048306
View Documents
Thermo Scientific Chemicals
B2048306
5 g
Each for $46.42
Only null left
 
AAB2048314
View Documents
Thermo Scientific Chemicals
B2048314
25 g
Each for $129.92
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5392-12-1
MFCD00004004

Specifications

5392-12-1
204°C to 206°C (18 mmHg)
MFCD00004004
1868308
3-phenylphthalide, 3-phenyl phthalide, phthalide, 3-phenyl, 3-phenyl-2-benzofuran-1 3h-one, 3-phenyl-1 3h-isobenzofuranone, 3-phenyl-1,3-dihydro-2-benzofuran-1-one, 1 3h-isobenzofuranone, 3-phenyl, 3-phenyl-3-hydroisobenzofuran-1-one, phenylphthalid, acmc-1ak9n
C1=CC=C(C=C1)C2C3=CC=CC=C3C(=O)O2
210.23
186.21
2-Methoxy-1-naphthaldehyde
81°C to 85°C
C14H10O2
5 g
Air Sensitive
SQFMIHCARVMICF-UHFFFAOYNA-N
3-phenyl-3H-2-benzofuran-1-one
95365
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 226-392-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only