missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-1-naphthaldehyde, 99%

Catalog No. aab2048314
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAB2048314 25 g
AAB2048306 5 g
2 options

Catalog No. AAB2048314

Supplier: Thermo Scientific Chemicals B2048314

Only null left

CAS: 5392-12-1 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5392-12-1
MDL Number MFCD00004004

Specifications

Melting Point 81°C to 85°C
Boiling Point 204°C to 206°C (18 mmHg)
Molecular Formula C12H10O2
Quantity 25 g
Beilstein 1868308
Sensitivity Air Sensitive
Synonym 2-methoxy-1-naphthaldehyde, 1-naphthalenecarboxaldehyde, 2-methoxy, 2-methoxy-1-naphthalenecarboxaldehyde, unii-7q7tx7tb9e, 7q7tx7tb9e, 2-methoxy-1-naphthylaldehyde, 2-methoxy-naphthalene-1-carbaldehyde, 2-methoxynaphthalenecarbaldehyde, 2-methoxy-1-naphthaldehyd, 2-methoxynaphthaldehyde
InChI Key YIQGLTKAOHRZOL-UHFFFAOYSA-N
SMILES COC1=C(C2=CC=CC=C2C=C1)C=O
IUPAC Name 2-methoxynaphthalene-1-carbaldehyde
Molecular Weight (g/mol) 186.21
PubChem CID 79352
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Methoxy-1-naphthaldehyde
Show More Show Less
EINECSNumber 226-392-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only