missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methacryloyloxy-2-methyladamantane (stabilized with MEHQ) 97.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 1 g |
| Formula Weight | 234.34 |
| Percent Purity | ≥97.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Methacryloyloxy-2-methyladamantane (stabilized with MEHQ) |
Safety and Handling
| EINECSNumber | (4)-1884 |
| TSCA | No |
| Recommended Storage | Freezer |