missing translation for 'onlineSavingsMsg'
Learn More

2-Mesitylenesulfonyl Hydrazide 97.0+%, TCI America™

Catalog No. M11985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11985G 5 g
M119825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11985G Supplier TCI America Supplier No. M11985G

Chemical Identifiers

CAS 16182-15-3
Molecular Formula C9H14N2O2S
Molecular Weight (g/mol) 214.283
MDL Number MFCD00007585
InChI Key JQUBKTQDNVZHIY-UHFFFAOYSA-N
Synonym 2,4,6-trimethylbenzenesulfonyl hydrazide, mesitylene-2-sulphonohydrazide, 2-mesitylenesulfonyl hydrazide, 2-mesitylenesulfonohydrazide, 2-mesitylenesulfono hydrazide, benzenesulfonic acid,2,4,6-trimethyl-, hydrazide, hydrazino 2,4,6-trimethylphenyl sulfone, mesitylenesulphonohydrazide, mesitylenesulfonylhydrazide, 2-mesitylehesulfonohydrazide
PubChem CID 85321
IUPAC Name 2,4,6-trimethylbenzenesulfonohydrazide
SMILES CC1=CC(=C(C(=C1)C)S(=O)(=O)NN)C

Specifications

Melting Point 114°C
Color White
Quantity 5 g
UN Number 1325
Formula Weight 214.28
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mesitylenesulfonyl Hydrazide