missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptothiazoline 98.0+%, TCI America™

Catalog No. p-7131963
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
500 g
Catalog No. Quantity
M006525G 25 g
M0065500G 500 g
2 options

Catalog No. M006525G

Supplier: TCI America M006525G

Only null left

Chemical Identifiers

CAS 96-53-7
Molecular Formula C3H5NS2
Molecular Weight (g/mol) 119.2
MDL Number MFCD00126013
InChI Key WGJCBBASTRWVJL-UHFFFAOYSA-N
Synonym 2-mercaptothiazoline, 2-thiazolidinethione, 2-thiazoline-2-thiol, thiazolidine-2-thione, metabasal, thyroidan, 2-mercapto-2-thiazoline, mercaptothiazoline, 2-thiothiazolidone, 2-thiazolidenethione
PubChem CID 2723699
IUPAC Name 1,3-thiazolidine-2-thione
SMILES C1CSC(=S)N1

Specifications

Melting Point 105°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 119.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptothiazoline
EINECSNumber (5)-0216
RTECSNumber XJ6122000
TSCA Yes