accessibility menu, dialog, popup

UserName

CAS RN 96-53-7

IUPAC Name: 1,3-thiazolidine-2-thione
Synonyms: 2-mercaptothiazoline
Molecular Weight (g/mol): 119.2
Molecular Formula: C3H5NS2
InChi Key: WGJCBBASTRWVJL-UHFFFAOYSA-N
SMILES: S=C1NCCS1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)