Learn More
2-Mercaptopyridine, 98%
CAS: 2637-34-5 | C5H5NS | 111.162 g/mol
$62.32 - $522.53
Chemical Identifiers
| CAS | 2637-34-5 |
|---|---|
| Molecular Formula | C5H5NS |
| Molecular Weight (g/mol) | 111.162 |
| MDL Number | MFCD00006285 |
| InChI Key | WHMDPDGBKYUEMW-UHFFFAOYSA-N |
| Synonym | 2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine |
| PubChem CID | 2723698 |
| ChEBI | CHEBI:45223 |
| IUPAC Name | 1H-pyridine-2-thione |
| SMILES | C1=CC(=S)NC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1174106
|
Thermo Scientific Chemicals
A1174106 |
5 g |
Each for $62.32
|
|
|||||
|
AAA1174114
|
Thermo Scientific Chemicals
A1174114 |
25 g |
Each for $183.59
|
|
|||||
|
AAA1174122
|
Thermo Scientific Chemicals
A1174122 |
100 g |
Each for $522.53
|
|
|||||
Description
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEmployed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.
Solubility
Slightly soluble in chloroform, methanol and dimethyl sulfoxide.
Notes
Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
| 2637-34-5 | |
| 111.162 | |
| WHMDPDGBKYUEMW-UHFFFAOYSA-N | |
| 2723698 | |
| 1H-pyridine-2-thione |
| C5H5NS | |
| MFCD00006285 | |
| 2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine | |
| CHEBI:45223 | |
| C1=CC(=S)NC=C1 |
Specifications
| 2637-34-5 | |
| C5H5NS | |
| 5 g | |
| Air sensitive | |
| Slightly soluble in chloroform,methanol and dimethyl sulfoxide. | |
| C1=CC(=S)NC=C1 | |
| 111.162 | |
| CHEBI:45223 | |
| 98% |
| 124°C to 130°C | |
| MFCD00006285 | |
| 105758 | |
| 2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine | |
| WHMDPDGBKYUEMW-UHFFFAOYSA-N | |
| 1H-pyridine-2-thione | |
| 2723698 | |
| 111.17 | |
| 2-Mercaptopyridine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 220-131-9
RTECSNumber : UT9600000
TSCA : Yes
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only