missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine, 98%

Catalog No. AAA1174106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1174106 5 g
AAA1174114 25 g
AAA1174122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1174106 Supplier Thermo Scientific Chemicals Supplier No. A1174106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2637-34-5 | C5H5NS | 111.162 g/mol

Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as a ligand in metal complexes. It can be used to coat porous media to purify plasmid DNA.

Solubility
Slightly soluble in chloroform, methanol and dimethyl sulfoxide.

Notes
Air Sensitive, keep the container tightly sealed. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 2637-34-5
Molecular Formula C5H5NS
Molecular Weight (g/mol) 111.162
MDL Number MFCD00006285
InChI Key WHMDPDGBKYUEMW-UHFFFAOYSA-N
Synonym 2-mercaptopyridine, 2-pyridinethiol, pyridine-2-thiol, 2-thiopyridine, pyrid-2-thione, 2 1h-pyridinethione, pyridinethione, 2-thiopyridone, 2-pyridinethione, 2-mercapto pyridine
PubChem CID 2723698
ChEBI CHEBI:45223
IUPAC Name 1H-pyridine-2-thione
SMILES C1=CC(=S)NC=C1

Specifications

Melting Point 124°C to 130°C
Quantity 5 g
Beilstein 105758
Sensitivity Air sensitive
Solubility Information Slightly soluble in chloroform,methanol and dimethyl sulfoxide.
Formula Weight 111.17
Percent Purity 98%
Chemical Name or Material 2-Mercaptopyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-131-9
RTECSNumber UT9600000
TSCA Yes
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only