Learn More
2-Mercaptoethylamine hydrochloride, 97+%
CAS: 156-57-0 | C2H8ClNS | 113.60 g/mol
Supplier: Thermo Scientific Chemicals A1437714
Description
An inhibitor of DMBA-induced mammary tumors2-Mercaptoethylamine hydrochloride acts as a precursor in taurine biosynthesis and component of coenzyme A. It is involved in the preparation of active pharmaceutical ingredients like ranitidine and nizatidine. It acts as an antidote for acetaminophen poisoning. Further, it is used in the oral treatment of nephropathic cystinosis, radiation sickness and disorders of cysteine excretion. In addition to this, it serves as an antioxidant and an inhibitor of 7,12-Dimethylbenz[a]anthracene (DMBA)-induced tumors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAn inhibitor of DMBA-induced mammary tumors2-Mercaptoethylamine hydrochloride acts as a precursor in taurine biosynthesis and component of coenzyme A. It is involved in the preparation of active pharmaceutical ingredients like ranitidine and nizatidine. It acts as an antidote for acetaminophen poisoning. Further, it is used in the oral treatment of nephropathic cystinosis, radiation sickness and disorders of cysteine excretion. In addition to this, it serves as an antioxidant and an inhibitor of 7,12-Dimethylbenz[a]anthracene (DMBA)-induced tumors.
Solubility
Soluble in water, ethanol, methanol and dimethyl sulfoxide.
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
| 2-Mercaptoethylamine hydrochloride | |
| 65°C to 69°C | |
| MFCD00012904 | |
| 3590083 | |
| 14,2779 | |
| Soluble in water,ethanol,methanol and dimethyl sulfoxide. | |
| [H+].[Cl-].NCCS | |
| 113.60 | |
| 113.61 |
| 156-57-0 | |
| C2H8ClNS | |
| 25 g | |
| Hygroscopic | |
| cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride | |
| OGMADIBCHLQMIP-UHFFFAOYSA-N | |
| 2-aminoethanethiol;hydrochloride | |
| 9082 | |
| ≥98% |
Chemical Identifiers
| 156-57-0 | |
| 113.60 | |
| OGMADIBCHLQMIP-UHFFFAOYSA-N | |
| 9082 | |
| [H+].[Cl-].NCCS |
| C2H8ClNS | |
| MFCD00012904 | |
| cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride | |
| 2-aminoethanethiol;hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 205-858-1
RTECSNumber : KJ0200000
TSCA : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only