accessibility menu, dialog, popup

UserName

CAS RN 156-57-0

IUPAC Name: hydrogen 2-aminoethane-1-thiol chloride
Synonyms: cysteamine hcl
Molecular Weight (g/mol): 113.6
Molecular Formula: C2H8ClNS
InChi Key: OGMADIBCHLQMIP-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCCS
Percent Purity 
  • (3)
  • (3)
  • (4)
Quantity 
  • (3)
  • (3)
  • (1)
  • (3)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (7)
  • (3)
  • (3)
  • (4)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)
  • (4)