Learn More
2-Mercaptoethylamine hydrochloride, 97+%
Description
An inhibitor of DMBA-induced mammary tumors2-Mercaptoethylamine hydrochloride acts as a precursor in taurine biosynthesis and component of coenzyme A. It is involved in the preparation of active pharmaceutical ingredients like ranitidine and nizatidine. It acts as an antidote for acetaminophen poisoning. Further, it is used in the oral treatment of nephropathic cystinosis, radiation sickness and disorders of cysteine excretion. In addition to this, it serves as an antioxidant and an inhibitor of 7,12-Dimethylbenz[a]anthracene (DMBA)-induced tumors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
An inhibitor of DMBA-induced mammary tumors2-Mercaptoethylamine hydrochloride acts as a precursor in taurine biosynthesis and component of coenzyme A. It is involved in the preparation of active pharmaceutical ingredients like ranitidine and nizatidine. It acts as an antidote for acetaminophen poisoning. Further, it is used in the oral treatment of nephropathic cystinosis, radiation sickness and disorders of cysteine excretion. In addition to this, it serves as an antioxidant and an inhibitor of 7,12-Dimethylbenz[a]anthracene (DMBA)-induced tumors.
Solubility
Soluble in water, ethanol, methanol and dimethyl sulfoxide.
Notes
Store in a cool place. Hygroscopic. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 65°C to 69°C |
| Quantity | 100 g |
| Beilstein | 3590083 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,2779 |
| Solubility Information | Soluble in water,ethanol,methanol and dimethyl sulfoxide. |
| Formula Weight | 113.61 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 2-Mercaptoethylamine hydrochloride |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 205-858-1 |
| RTECSNumber | KJ0200000 |
| TSCA | Yes |
| Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only