missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethyl sulfide, 90%, Thermo Scientific™

Chemical Identifiers

CAS 3570-55-6
Molecular Formula C4H10S3
Molecular Weight (g/mol) 154.30
MDL Number MFCD00004893
InChI Key KSJBMDCFYZKAFH-UHFFFAOYSA-N
Synonym 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
PubChem CID 77117
IUPAC Name 2-(2-sulfanylethylsulfanyl)ethanethiol
SMILES SCCSCCS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC125490100
View Documents Promotion Details
N/A 10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC125490500
View Documents Promotion Details
N/A 50g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

3570-55-6
154.30
KSJBMDCFYZKAFH-UHFFFAOYSA-N
77117
SCCSCCS
C4H10S3
MFCD00004893
2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
2-(2-sulfanylethylsulfanyl)ethanethiol

Specifications

3570-55-6
135.0°C to 136.0°C (10.0mmHg)
Authentic
C4H10S3
(HSCH2CH2)2S
1.183
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,toluene,chloroform,,dioxane,carbon tetrachloride
SCCSCCS
154.30
154.32
2-Mercaptoethyl sulfide
1.1830g/mL
90°C
88 % min. (GC)
1.594
MFCD00004893
2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
KSJBMDCFYZKAFH-UHFFFAOYSA-N
2-(2-sulfanylethylsulfanyl)ethanethiol
77117
90%
Videos
Safety and Handling

Safety and Handling

Warning

EINECSNumber : 222-671-

SDS
Documents

Documents

RUO – Research Use Only