missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Mercaptoethyl)pyrazine 98.0+%, TCI America™
$70.84 - $210.63
Chemical Identifiers
| CAS | 35250-53-4 |
|---|---|
| Molecular Formula | C6H8N2S |
| Molecular Weight (g/mol) | 140.204 |
| MDL Number | MFCD00053160 |
| InChI Key | QKVWBAMZPUHCMO-UHFFFAOYSA-N |
| Synonym | pyrazineethanethiol, 2-pyrazinylethanethiol, 2-pyrazineethanethiol, pyrazine ethanethiol, pyrazinyl ethanethiol, 2-2-mercaptoethyl pyrazine, 2-pyrazinylethylmercaptan, 2-mercaptoethyl pyrazine, mercaptoethylpyrazine, unii-f7i6g0e56f |
| PubChem CID | 61945 |
| IUPAC Name | 2-pyrazin-2-ylethanethiol |
| SMILES | C1=CN=C(C=N1)CCS |
Chemical Identifiers
| 35250-53-4 | |
| 140.204 | |
| QKVWBAMZPUHCMO-UHFFFAOYSA-N | |
| 61945 | |
| C1=CN=C(C=N1)CCS |
| C6H8N2S | |
| MFCD00053160 | |
| pyrazineethanethiol, 2-pyrazinylethanethiol, 2-pyrazineethanethiol, pyrazine ethanethiol, pyrazinyl ethanethiol, 2-2-mercaptoethyl pyrazine, 2-pyrazinylethylmercaptan, 2-mercaptoethyl pyrazine, mercaptoethylpyrazine, unii-f7i6g0e56f | |
| 2-pyrazin-2-ylethanethiol |
Specifications
| 35250-53-4 | |
| 110°C | |
| MFCD00053160 | |
| 2810 | |
| QKVWBAMZPUHCMO-UHFFFAOYSA-N | |
| 2-pyrazin-2-ylethanethiol | |
| 61945 | |
| ≥98.0% (GC) | |
| (2-Mercaptoethyl)pyrazine |
| Yellow | |
| C6H8N2S | |
| 1 g | |
| pyrazineethanethiol, 2-pyrazinylethanethiol, 2-pyrazineethanethiol, pyrazine ethanethiol, pyrazinyl ethanethiol, 2-2-mercaptoethyl pyrazine, 2-pyrazinylethylmercaptan, 2-mercaptoethyl pyrazine, mercaptoethylpyrazine, unii-f7i6g0e56f | |
| C1=CN=C(C=N1)CCS | |
| 140.204 | |
| 140.20 | |
| Liquid |
Safety and Handling
RTECSNumber : KJ2551000
TSCA : Yes
Recommended Storage : Refrigerator