accessibility menu, dialog, popup

UserName

CAS RN 35250-53-4

IUPAC Name: 2-(pyrazin-2-yl)ethane-1-thiol
Synonyms: pyrazineethanethiol
Molecular Weight (g/mol): 140.2
Molecular Formula: C6H8N2S
InChi Key: QKVWBAMZPUHCMO-UHFFFAOYSA-N
SMILES: SCCC1=CN=CC=N1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results