missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Mercaptoethyl)pyrazine 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 110°C |
| Quantity | 5 g |
| UN Number | 2810 |
| Formula Weight | 140.20 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | (2-Mercaptoethyl)pyrazine |
Safety and Handling
| RTECSNumber | KJ2551000 |
| TSCA | Yes |
| Recommended Storage | Refrigerator |